Anti-PHYHIPL Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9719A
89398-974EA 436.86 USD
89398-974
Anti-PHYHIPL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PHYHIPL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR