Anti-PSMG2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9698B
89398-928EA 538.45 USD
89398-928
Anti-PSMG2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSMG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR