Anti-CA8 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9692B
89398-916EA 476.18 USD
89398-916
Anti-CA8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CA8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR