Anti-DGAT2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9686A
89398-904EA 436.86 USD
89398-904
Anti-DGAT2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DGAT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR