Anti-GSPT2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9663C
89398-858EA 610.01 USD
89398-858
Anti-GSPT2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GSPT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR