Anti-TMUB2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9642C
89398-814EA 610.01 USD
89398-814
Anti-TMUB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TMUB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR