Anti-CENPI Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9618C
89398-768EA 610.01 USD
89398-768
Anti-CENPI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPI
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR