Anti-COX6A2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9616C
89398-764EA 436.86 USD
89398-764
Anti-COX6A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX6A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR