Anti-SLCO1B1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9609C
89398-750EA 436.77 USD
89398-750
Anti-SLCO1B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLCO1B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR