Anti-CK2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9594A
89398-716EA 580.97 USD
89398-716
Anti-CK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR