Anti-CDC16 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9572A
89398-674EA 436.73 USD
89398-674
Anti-CDC16 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC16
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR