Anti-RPL37 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9565B
89398-660EA 610.34 USD
89398-660
Anti-RPL37 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPL37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR