Anti-LRRC4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9530B
89398-592EA 610.34 USD
89398-592
Anti-LRRC4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LRRC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR