Anti-NUCKS1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9527A
89398-586EA 610.01 USD
89398-586
Anti-NUCKS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NUCKS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR