Anti-IL10RB Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9465C
89398-464EA 580.8 USD
89398-464
Anti-IL10RB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL10RB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR