Anti-A1BG Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9444C
89398-418EA 610.01 USD
89398-418
Anti-A1BG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: A1BG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR