Anti-SLC39A8 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9359C
89398-242EA 610.01 USD
89398-242
Anti-SLC39A8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC39A8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR