Anti-DTNB Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9187B
89397-894EA 580.97 USD
89397-894
Anti-DTNB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DTNB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR