Anti-COX5A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9154C
89397-824EA 610.01 USD
89397-824
Anti-COX5A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR