Anti-CDK5RAP1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9034B
89397-580EA 610.01 USD
89397-580
Anti-CDK5RAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK5RAP1
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR