Anti-FOXC1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8907B
89397-334EA 609.84 USD
89397-334
Anti-FOXC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOXC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR