Anti-CYP51A1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8874C
89397-266EA 610.34 USD
89397-266
Anti-CYP51A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP51A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR