Anti-ATF7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8844A
89397-206EA 610.17 USD
89397-206
Anti-ATF7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATF7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR