Anti-CST7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8824A
89397-168EA 580.97 USD
89397-168
Anti-CST7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CST7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR