Anti-HIST1H2AH Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8820A
89397-160EA 610.01 USD
89397-160
Anti-HIST1H2AH Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIST1H2AH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR