Anti-CYP8B1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8787B
89397-096EA 609.84 USD
89397-096
Anti-CYP8B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP8B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR