Anti-DERA Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8764A
89397-048EA 610.01 USD
89397-048
Anti-DERA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DERA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR