Anti-CYP2A6 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8724A
89396-970EA 580.97 USD
89396-970
Anti-CYP2A6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP2A6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR