Anti-CYP11B1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8723A
89396-966EA 580.97 USD
89396-966
Anti-CYP11B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP11B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR