Anti-BMPR1A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8695A
89396-908EA 580.97 USD
89396-908
Anti-BMPR1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BMPR1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR