Anti-CHL1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8662C
89396-840EA 580.97 USD
89396-840
Anti-CHL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR