Anti-MFRP Rabbit Polyclonal Antibody

Supplier: Abcepta
AP8661B
89396-838EA 580.8 USD
89396-838
Anti-MFRP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MFRP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR