Anti-SDHAF1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP5392C
89389-516EA 610.01 USD
89389-516
Anti-SDHAF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SDHAF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR