Anti-HIF1A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP4776A
89388-282EA 617.76 USD
89388-282
Anti-HIF1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR