Anti-CDK2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP3613A
89387-848EA 610.34 USD
89387-848
Anti-CDK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR