Anti-PRKDC Rabbit Polyclonal Antibody

Supplier: Abcepta
AP3483A
89387-598EA 581.28 USD
89387-598
Anti-PRKDC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKDC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR