Anti-MAPK8 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP3427A
89387-488EA 581.28 USD
89387-488
Anti-MAPK8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR