Anti-PGLS Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2935B
89386-854EA 609.84 USD
89386-854
Anti-PGLS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PGLS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR