Anti-TUFM Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2918A
89386-802EA 610.34 USD
89386-802
Anti-TUFM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TUFM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR