Anti-CD38 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP2878B
89386-680EA 610.34 USD
89386-680
Anti-CD38 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR