Anti-RASAL Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12460C
89381-312EA 610.01 USD
89381-312
Anti-RASAL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RASAL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR