Anti-PDP1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12453C
89381-296EA 610.34 USD
89381-296
Anti-PDP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR