Anti-TFR2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12449B
89381-286EA 580.97 USD
89381-286
Anti-TFR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TFR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR