Anti-PIGA Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12378B
89381-136EA 609.84 USD
89381-136
Anti-PIGA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIGA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR