Anti-OR52N5 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12368B
89381-116EA 610.01 USD
89381-116
Anti-OR52N5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OR52N5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR