Anti-COX6B1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12336B
89381-046EA 610.01 USD
89381-046
Anti-COX6B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COX6B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR