Anti-PDE6B Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12294B
89380-952EA 580.97 USD
89380-952
Anti-PDE6B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDE6B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR