Anti-CD44 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12156B
89380-638EA 580.97 USD
89380-638
Anti-CD44 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR