Anti-AP1M1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12036C
89380-400EA 610.34 USD
89380-400
Anti-AP1M1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AP1M1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR