Anti-PCDHB15 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12017A
89380-360EA 610.01 USD
89380-360
Anti-PCDHB15 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PCDHB15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR